Search
Search

Ddx54| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028041.2 | NP_082317.1 | 6 - 7 | 659 | 96 | |
| GenBank mRNA | AF319547.1 | - | 6 - 7 | 1019 | 96 | |
| AK010484.1 | - | 3 - 4 | 239 | 96 | ||
| AK028274.1 | - | 6 - 7 | 673 | 96 | ||
| AK031534.1 | - | 6 - 7 | 669 | 96 | ||
| AK031679.1 | - | 6 - 7 | 677 | 96 | ||
| AK147715.1 | - | 6 - 7 | 674 | 96 | ||
| AK152989.1 | - | 6 - 7 | 673 | 97 | ||
| AK155138.1 | - | 6 - 7 | 671 | 96 | ||
| AK167603.1 | - | 6 - 7 | 673 | 96 | ||
| AK168455.1 | - | 6 - 7 | 673 | 96 | ||
| AK169255.1 | - | 6 - 7 | 672 | 96 | ||
| AK170967.1 | - | 6 - 7 | 671 | 96 | ||
| AK212480.1 | - | 2 - 3 | 86 | 96 | ||
| BC043699.1 | - | 6 - 7 | 659 | 5344158 | 96 | |
| BC055026.1 | - | 2 - 3 | 102 | 96 | ||
| BC062920.1 | - | 6 - 7 | 662 | 96 | ||
Set Membership: |
|