Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203755_g1
          See other MOSPD3 GE Assays ›
          Gene Symbol:
          Mospd3
          Gene Name:
          motile sperm domain containing 3
          Gene Aliases:
          1190005J19Rik, 5133401H10Rik, Gtig2, R124
          Chromosome Location:
          Chr.5: 137596645 - 137601083 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203755_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mospd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001254762.1 NP_001241691.1 4 - 5 907 63
          NM_030037.2 NP_084313.1 5 - 6 1060 63
          GenBank mRNA AK088731.1 - 4 - 5 898 63
          BC003880.1 - 4 - 5 917 3496491 63
          BC034167.1 - 5 - 6 1045 63
          BC039620.1 - 5 - 6 1041 5051528 63
          BQ445267.1 - 3 - 4 473 63
          DV656015.1 - 2 - 3 317 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mospd3

          Gene Ontology Categories:

          Function(s) Process(es)

          heart development
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126543 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126537 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126539 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126546 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126544 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126538 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978676 BC034167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978676 BC039620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978676 AK004495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978676 NM_030037.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978676 AK088731.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177614 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101318 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978676 BC034167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978676 BC039620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978676 AK004495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978676 NM_030037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978676 AK088731.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492228 BC034167.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492228 BC039620.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492228 NM_030037.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492228 BC003880.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P62051 BC034167.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P62051 BC039620.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P62051 NM_030037.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P62051 AK088731.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P62051 BC003880.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126531 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05761005 - -
          Whole Mouse Genome G4122A A_52_P62051 BC034167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62051 BC039620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62051 NM_030037.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62051 AK088731.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P62051 BC003880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492228 BC034167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492228 BC039620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492228 NM_030037.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492228 BC003880.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131098 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04460835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04460840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04385730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04460830 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04460838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126543 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126537 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126542 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126546 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126544 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04683368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04181587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04460835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04127201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04303710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04517098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04460832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04792998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04642059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04460845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04361435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04800679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04158820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04460830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04615761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04517099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04514068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04460843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534857 - -
          Affymetrix HT MG-430 PM Array Plate 1460452_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453961_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534854 - -
          Mouse Genome 430 2.0 Array 1453961_PM_a_at - -
          Mouse Genome 430 2.0 Array 1453961_a_at - -
          Mouse Genome 430 2.0 Array 1460452_PM_at - -
          Mouse Genome 430 2.0 Array 1460452_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68929_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68929_at - -
          Affymetrix Mouse Clariom S Array PSR0500031407.mm.1 NM_001254762.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031407.mm.1 NM_001254762.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031407.mm.1 NM_030037.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031407.mm.1 TC0500003449.mm.1 MTA TC
          Mouse Expression Set 430 1453961_PM_a_at - -
          Mouse Expression Set 430 1460452_at - -
          Mouse Expression Set 430 1460452_PM_at - -
          Mouse Expression Set 430 1453961_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 835839 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 540242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677531 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677531 - -
          mouse-6 v1.1 (Illumina) scl25876.6.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 540242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25876.6.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3153772 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677531 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677531 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3153772 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline