Search
Search

Prmt3| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133740.2 | NP_598501.1 | 10 - 11 | 1101 | 82 | |
| GenBank mRNA | AK010534.1 | - | 10 - 11 | 1113 | 82 | |
| AK031738.1 | - | 10 - 11 | 1102 | 82 | ||
| AK086646.1 | - | 10 - 11 | 1060 | 82 | ||
| AK155236.1 | - | 10 - 11 | 1099 | 82 | ||
| BC008128.1 | - | 6 - 7 | 644 | 82 | ||
| BC050775.1 | - | 10 - 11 | 1080 | 6534023 | 82 | |
Set Membership: |
|