Search
Search

Saal1| Gene Symbol: Saal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030233.1 | NP_084509.1 | 1 - 2 | 153 | 76 | |
| GenBank mRNA | AB489137.1 | - | 1 - 2 | 134 | 76 | |
| AK019878.1 | - | 1 - 2 | 154 | 76 | ||
| AK030833.1 | - | 1 - 2 | 159 | 76 | ||
| AK033202.1 | - | 1 - 2 | 164 | 76 | ||
| AK035874.1 | - | 1 - 2 | 164 | 76 | ||
| AK132193.1 | - | 1 - 2 | 154 | 76 | ||
| AK134418.1 | - | 1 - 2 | 161 | 76 | ||
| BC019985.1 | - | 1 - 2 | 147 | 76 | ||
| BC068127.1 | - | 1 - 2 | 150 | 76 | ||
| BC087944.1 | - | 1 - 2 | 165 | 30314036 | 76 | |
Set Membership: |
|