Search
Search

Cldn15| Gene Symbol: Cldn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021719.4 | NP_068365.1 | 3 - 4 | 695 | 76 | |
| GenBank mRNA | AK008227.1 | - | 3 - 4 | 493 | 76 | |
| AK008683.1 | - | 3 - 4 | 696 | 76 | ||
| BC023428.1 | - | 3 - 4 | 684 | 5250004 | 76 | |
Set Membership: |
|