Search
Search

Ap2a1| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077264.1 | NP_001070732.1 | 4 - 5 | 615 | 70 | |
| NM_007458.2 | NP_031484.1 | 4 - 5 | 615 | 70 | ||
| GenBank mRNA | AK135424.1 | - | 4 - 5 | 544 | 70 | |
| AK155515.1 | - | 4 - 5 | 551 | 70 | ||
| BC031433.1 | - | 4 - 5 | 565 | 4481643 | 70 | |
| CA327188.1 | - | 4 - 5 | 621 | 70 | ||
Set Membership: |
|