Search
Search

Slc17a7| Gene Symbol: Slc17a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182993.2 | NP_892038.2 | 6 - 7 | 888 | 75 | |
| GenBank mRNA | AK159064.1 | - | 6 - 7 | 861 | 75 | |
| BC028938.1 | - | 4 - 5 | 327 | 75 | ||
| BC054462.1 | - | 4 - 5 | 670 | 6494471 | 75 | |
Set Membership: |
|