Search
Search

Crybb1| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023695.2 | NP_076184.1 | 1 - 2 | 32 | 86 | |
| GenBank mRNA | AF106853.1 | - | 1 - 2 | 32 | 86 | |
| AK014012.1 | - | 1 - 2 | 34 | 86 | ||
| AK053869.1 | - | 1 - 2 | 33 | 86 | ||
Set Membership: |
|