Search
Search

Tpst2| Gene Symbol: Tpst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009419.3 | NP_033445.2 | 1 - 2 | 66 | 69 | |
| GenBank mRNA | AK075672.1 | - | 1 - 2 | 66 | 69 | |
| AK149379.1 | - | 1 - 2 | 70 | 69 | ||
| BC003721.1 | - | 1 - 2 | 57 | 3494892 | 69 | |
Set Membership: |
|