Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207130_g1
          See other TSC22D4 GE Assays ›
          Gene Symbol:
          Tsc22d4
          Gene Name:
          TSC22 domain family, member 4
          Gene Aliases:
          0610009M14Rik, 1700023B23Rik, AI415410, Thg-1pit, Tilz2
          Chromosome Location:
          Chr.5: 137745969 - 137768453 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207130_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsc22d4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023910.6 NP_076399.4 1 - 2 442 137
          NM_029805.1 NP_084081.1 1 - 2 442 137
          GenBank mRNA AF201286.1 - 1 - 2 154 137
          AK018735.2 - 1 - 2 415 137
          AK040534.1 - 1 - 2 379 137
          AK079510.1 - 1 - 2 168 137
          AK080828.1 - 1 - 2 453 137
          AK143608.1 - 1 - 2 219 137
          AK150043.1 - 1 - 2 413 137
          AK154153.1 - 1 - 2 392 137
          AK155712.1 - 1 - 2 443 137
          AK170487.1 - 1 - 2 431 137
          BC023761.1 - 1 - 2 346 5343927 137
          BC145721.1 - 1 - 2 405 40131024 137
          BC145723.1 - 1 - 2 405 40131026 137
          BY750078.1 - 1 - 2 442 137

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF201286.1, AK018735.2, AK040534.1, AK079510.1, AK143608.1, AK154153.1, AK155712.1, AK170487.1, BC023761.1, BC145721.1, BC145723.1, NM_023910.6, NM_029805.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsc22d4

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          response to osmotic stress
          transcription factor activity, sequence-specific DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500014899.mm.1 NM_023910.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014899.mm.1 NM_023910.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014899.mm.1 NM_029805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014899.mm.1 NM_029805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014899.mm.1 TC0500001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014903.mm.1 NM_023910.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014903.mm.1 NM_023910.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014903.mm.1 NM_029805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014903.mm.1 NM_029805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014903.mm.1 TC0500001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014900.mm.1 NM_023910.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014900.mm.1 NM_023910.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014900.mm.1 NM_029805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014900.mm.1 NM_029805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014900.mm.1 TC0500001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014902.mm.1 NM_023910.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014902.mm.1 NM_023910.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014902.mm.1 NM_029805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014902.mm.1 NM_029805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014902.mm.1 TC0500001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014901.mm.1 NM_023910.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014901.mm.1 NM_023910.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014901.mm.1 NM_029805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014901.mm.1 NM_029805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014901.mm.1 TC0500001634.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014896.mm.1 NM_023910.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014896.mm.1 NM_023910.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014896.mm.1 NM_029805.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014896.mm.1 NM_029805.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014896.mm.1 TC0500001634.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664037 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline