Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207417_gH
          See other REN2 GE Assays ›
          Gene Symbol:
          Ren2
          Gene Name:
          renin 2 tandem duplication of Ren1
          Gene Aliases:
          Ren, Ren-2, Ren-B, Rn-2, Rnr
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207417_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ren2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031193.2 NP_112470.2 7 - 8 1090 88
          GenBank mRNA BC011157.1 - 7 - 8 966 4190969 88
          BG867251.1 - 1 - 2 128 88
          J00621.1 - 7 - 8 990 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001206.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ren2

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          positive regulation of blood pressure
          endopeptidase activity
          aspartic-type endopeptidase activity
          peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959091 NM_031193.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959091 BG867251.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730744 AK020290.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298623 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131022 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959091 NM_031193.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959091 BG867251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P192265 J00621.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00489500 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00489507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00489509 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10349923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349926 - -
          Affymetrix Mouse Gene 1.1 ST Array 10349922 - -
          Affymetrix Murine Genome 6500A Array Msa11330_at - -
          Affymetrix Mouse Clariom S Array PSR0100009896.mm.1 NM_031192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009896.mm.1 NM_031192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009896.mm.1 NM_031193.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009896.mm.1 TC0100001206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009897.mm.1 NM_031192.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009897.mm.1 NM_031192.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009897.mm.1 NM_031193.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009897.mm.1 TC0100001206.mm.1 MTA TC
          Mouse Expression Set 430 1448975_s_at - -
          Mouse Expression Set 430 1448975_PM_s_at - -
          Murine Genome U74v2 Set 161269_i_at - -
          Murine Genome U74v2 Set 98480_PM_s_at - -
          Murine Genome U74v2 Set 98480_s_at - -
          Murine Genome U74v2 Set 161269_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1448975_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1448975_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 585369 - -
          ABI Mouse Genome Survey Microarray 925906 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6380735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620114 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620114 - -
          mouse-6 v1.1 (Illumina) scl019702.11_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019702.11_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2777226 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777226 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019702.11_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline