Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207896_s1
          See other VMN1R87 GE Assays ›
          Gene Symbol:
          Vmn1r87
          Gene Name:
          vomeronasal 1 receptor 87
          Gene Aliases:
          V1rk1
          Chromosome Location:
          Chr.7: 13131471 - 13140087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207896_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134227.1 NP_598988.1 1 653 127

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r87

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088315 NM_134227.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144719 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088315 NM_134227.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P207054 NM_134227.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974618 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06126136 - -
          Whole Mouse Genome G4122A A_51_P207054 NM_134227.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108304 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06126136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05988787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974621 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560092 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560091 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 304139 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5360193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231428 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231428 - -
          mouse-6 v1.1 (Illumina) scl9195.1.1_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9195.1.1_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231428 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231428 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline