Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01207980_m1
          See other ZFP943 GE Assays ›
          Gene Symbol:
          Zfp943
          Gene Name:
          zinc finger prtoein 943
          Gene Aliases:
          4930432O21Rik
          Chromosome Location:
          Chr.17: 21962559 - 21994366 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01207980_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp943
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025373.2 NP_001020544.1 3 - 4 464 122
          GenBank mRNA BC066031.1 - 3 - 4 265 6418362 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000284.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp943

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31088171 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059055 BC066031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059055 BE447849.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059055 NM_001025373.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624338 AK049440.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624338 AK147117.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624338 BC006185.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624338 BC028921.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624338 M91380.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624338 NM_008047.5 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24160 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162842 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165991 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059055 BC066031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059055 BE447849.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059055 NM_001025373.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31088176 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735135 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143028 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735330 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735109 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03291279 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735143 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163821 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735083 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735288 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735064 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735123 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162842 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735362 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165991 - -
          Whole Mouse Genome G4122A A_52_P456296 AK015297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229359 BC066031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229359 NM_001025373.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114203 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03291279 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102487 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143028 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165991 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162842 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03291272 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08735143 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08735135 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08735064 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163821 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08735288 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13154323 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31088171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31087999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31087979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31087986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31087990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31087993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31088245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31087991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13538880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13291624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13154333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31087998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13072634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13291622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31087986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31087988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31087989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31087993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31087990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31088181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442260 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442261 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442259 - -
          Affymetrix HT MG-430 PM Array Plate 1430727_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434625_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442258 - -
          Mouse Genome 430 2.0 Array 1430727_PM_at - -
          Mouse Genome 430 2.0 Array 1434625_PM_at - -
          Mouse Genome 430 2.0 Array 1430727_at - -
          Mouse Genome 430 2.0 Array 1434625_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74670_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74670_at - -
          Affymetrix Mouse Clariom S Array PSR1700002109.mm.1 NM_001025373.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002109.mm.1 NM_001025373.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002109.mm.1 TC1700000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002108.mm.1 NM_001025373.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002108.mm.1 NM_001025373.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002108.mm.1 TC1700000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002114.mm.1 NM_001025373.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002114.mm.1 NM_001025373.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002114.mm.1 TC1700000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002111.mm.1 NM_001025373.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002111.mm.1 NM_001025373.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002111.mm.1 TC1700000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002112.mm.1 NM_001025373.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002112.mm.1 NM_001025373.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002112.mm.1 TC1700000284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002110.mm.1 NM_001025373.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002110.mm.1 NM_001025373.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002110.mm.1 TC1700000284.mm.1 MTA TC
          Mouse Expression Set 430 1430727_at - -
          Mouse Expression Set 430 1430727_PM_at - -
          Mouse Expression Set 430 1434625_PM_at - -
          Mouse Expression Set 430 1434625_at - -
          Murine Genome U74v2 Set 160725_at - -
          Murine Genome U74v2 Set 160725_PM_at - -
          Mouse Genome 430A 2.0 Array 1430727_at - -
          Mouse Genome 430A 2.0 Array 1430727_PM_at - -
          Mouse Genome 430A 2.0 Array 1434625_at - -
          Mouse Genome 430A 2.0 Array 1434625_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1430727_at - -
          Affymetrix HT Mouse Genome 430A Array 1434625_at - -
          Affymetrix HT Mouse Genome 430A Array 1430727_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434625_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3006178 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3006178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline