Search
Search

Cnot6l| Gene Symbol: Cnot6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285511.1 | NP_001272440.1 | 3 - 4 | 496 | 71 | |
| NM_001285514.1 | NP_001272443.1 | 3 - 4 | 370 | 71 | ||
| NM_144910.2 | NP_659159.1 | 3 - 4 | 364 | 71 | ||
| NM_178854.4 | NP_849185.2 | 3 - 4 | 374 | 71 | ||
| GenBank mRNA | AK030112.1 | - | 3 - 4 | 372 | 71 | |
| AK151728.1 | - | 3 - 4 | 499 | 71 | ||
| BC018506.1 | - | 3 - 4 | 364 | 3598248 | 71 | |
| CF173938.1 | - | 3 - 4 | 346 | 71 | ||
Set Membership: |
|