Search
Search
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048177.2 | NP_001041642.1 | 20 - 21 | 3224 | 170 | |
| NM_008413.3 | NP_032439.2 | 21 - 22 | 3307 | 170 | ||
| GenBank mRNA | BC054807.1 | - | 20 - 21 | 2987 | 6838318 | 170 |
| BC059834.1 | - | 20 - 21 | 2987 | 6838318 | 170 | |
| L16956.1 | - | 20 - 21 | 2976 | 170 | ||
Set Membership: |
|