Search
Search

Gnpda2| Gene Symbol: Gnpda2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038015.1 | NP_001033104.1 | 2 - 3 | 242 | 156 | |
| GenBank mRNA | AK014950.1 | - | 2 - 3 | 252 | 156 | |
| AK015527.1 | - | 2 - 3 | 252 | 156 | ||
| AK016785.1 | - | 297 | 156 | |||
| AK017588.1 | - | 2 - 3 | 251 | 156 | ||
| AK051596.1 | - | 2 - 3 | 251 | 156 | ||
| AK141408.1 | - | 2 - 3 | 254 | 156 | ||
| AK166601.1 | - | 2 - 3 | 244 | 156 | ||
| AK168516.1 | - | 2 - 3 | 261 | 156 | ||
| AK171299.1 | - | 2 - 3 | 237 | 156 | ||
| AK220275.1 | - | 2 - 3 | 203 | 156 | ||
| BC004084.1 | - | 2 - 3 | 242 | 3593102 | 156 | |
Set Membership: |
|