Search
Search

Glud1| Gene Symbol: Glud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008133.4 | NP_032159.1 | 6 - 7 | 1092 | 63 | |
| GenBank mRNA | AK075971.1 | - | 6 - 7 | 996 | 63 | |
| AK089152.1 | - | 6 - 7 | 995 | 63 | ||
| AK150637.1 | - | 6 - 7 | 1092 | 63 | ||
| AK161876.1 | - | 2 - 3 | 226 | 63 | ||
| BC052724.1 | - | 6 - 7 | 1066 | 6812055 | 63 | |
| BC057347.1 | - | 6 - 7 | 1066 | 6812055 | 63 | |
| X57024.1 | - | 6 - 7 | 997 | 63 | ||
Set Membership: |
|