| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300016036.mm.1 |
NM_026641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016036.mm.1 |
NM_026641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016036.mm.1 |
TC0300002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300016040.mm.1 |
NM_026641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016040.mm.1 |
NM_026641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016040.mm.1 |
TC0300002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300016030.mm.1 |
NM_026641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016030.mm.1 |
NM_026641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016030.mm.1 |
TC0300002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300016031.mm.1 |
NM_026641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016031.mm.1 |
NM_026641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016031.mm.1 |
TC0300002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300016024.mm.1 |
NM_026641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016024.mm.1 |
NM_026641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300016024.mm.1 |
TC0300002122.mm.1 |
MTA TC |