Search
Search

Gata5| Gene Symbol: Gata5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008093.2 | NP_032119.2 | 5 - 6 | 1368 | 60 | |
| GenBank mRNA | AK132020.1 | - | 5 - 6 | 1389 | 60 | |
| AK142213.1 | - | 5 - 6 | 1370 | 60 | ||
| BC105653.1 | - | 6 - 7 | 1081 | 40046623 | 60 | |
| BC105654.1 | - | 6 - 7 | 1142 | 40046622 | 60 | |
| BC105655.1 | - | 6 - 7 | 991 | 40046621 | 60 | |
| BC105656.1 | - | 6 - 7 | 776 | 60 | ||
| U84725.1 | - | 5 - 6 | 1362 | 60 | ||
Set Membership: |
|