Search
Search

C2cd2| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174847.2 | NP_777272.2 | 11 - 12 | 1738 | 67 | |
| GenBank mRNA | AK017896.1 | - | 11 - 12 | 1723 | 67 | |
| AK077731.1 | - | 11 - 12 | 1699 | 67 | ||
| AK080956.1 | - | 11 - 12 | 1682 | 67 | ||
| AK086520.1 | - | 11 - 12 | 1741 | 67 | ||
| AK143875.1 | - | 11 - 12 | 1712 | 67 | ||
| AK169902.1 | - | 11 - 12 | 1674 | 67 | ||
| AK196045.1 | - | 2 - 3 | 173 | 67 | ||
| AK220454.1 | - | 11 - 12 | 1723 | 67 | ||
| BC038064.1 | - | 1 - 2 | 170 | 67 | ||
| BC043688.1 | - | 11 - 12 | 1699 | 5136661 | 67 | |
Set Membership: |
|