Search
Search
| Gene Symbol: Flt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010229.2 | NP_034359.2 | 15 - 16 | 2167 | 107 | |
| GenBank mRNA | AK149292.1 | - | 15 - 16 | 1985 | 107 | |
| BC109003.1 | - | 15 - 16 | 1960 | 107 | ||
| BC109004.1 | - | 15 - 16 | 1972 | 107 | ||
| M64689.1 | - | 15 - 16 | 1973 | 107 | ||
| X59398.1 | - | 15 - 16 | 2024 | 107 | ||
Set Membership: |
|