Search
Search
| Gene Symbol: Yes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205132.1 | NP_001192061.1 | 10 - 11 | 1501 | 141 | |
| NM_001205133.1 | NP_001192062.1 | 10 - 11 | 1501 | 141 | ||
| NM_009535.3 | NP_033561.1 | 10 - 11 | 1963 | 141 | ||
| GenBank mRNA | AK052478.1 | - | 10 - 11 | 1453 | 141 | |
| AK167420.1 | - | 10 - 11 | 1638 | 141 | ||
| BC010594.1 | - | 10 - 11 | 1337 | 3989783 | 141 | |
| L25762.1 | - | 1 - 2 | 124 | 141 | ||
| X67677.1 | - | 10 - 11 | 1890 | 141 | ||
Set Membership: |
|