Search
Search

Lipo1| Gene Symbol: Lipo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013770.3 | NP_001013792.2 | 2 - 3 | 567 | 143 | |
| GenBank mRNA | AK139790.1 | - | 2 - 3 | 568 | 143 | |
| AK170332.1 | - | 2 - 3 | 501 | 143 | ||
Set Membership: |
|