Search
Search
| Gene Symbol: Rft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177815.3 | NP_808483.2 | 7 - 8 | 813 | 86 | |
| GenBank mRNA | AK184779.1 | - | 2 - 3 | 213 | 87 | |
| BC082539.1 | - | 7 - 8 | 781 | 86 | ||
| BC094336.1 | - | 7 - 8 | 781 | 86 | ||
| BC141275.1 | - | 7 - 8 | 807 | 9055887 | 86 | |
Set Membership: |
|