Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01212659_m1
          See other PHACTR3 GE Assays ›
          Gene Symbol:
          Phactr3
          Gene Name:
          phosphatase and actin regulator 3
          Gene Aliases:
          1500003N10Rik, 4930415A02Rik, H17739, Scapin1, Scapinin, mKIAA4224
          Chromosome Location:
          Chr.2: 178118975 - 178338492 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01212659_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phactr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007154.3 NP_001007155.1 11 - 12 1583 90
          NM_001177789.1 NP_001171260.1 11 - 12 1801 90
          NM_001177790.1 NP_001171261.1 11 - 12 1798 90
          NM_001177791.1 NP_001171262.1 11 - 12 1635 90
          NM_028806.2 NP_083082.1 11 - 12 2040 90
          GenBank mRNA AJ632220.1 - 11 - 12 1598 90
          AJ632221.1 - 11 - 12 1636 90
          AJ632222.1 - 11 - 12 1583 90
          AJ632223.1 - 10 - 11 1473 90
          AK005136.1 - 11 - 12 1605 90
          AK032242.1 - 11 - 12 1807 90
          AK038449.1 - 11 - 12 1551 90
          AK220554.1 - 11 - 12 1798 90
          AY501387.1 - 11 - 12 1747 90
          BC058621.1 - 11 - 12 1956 5708186 90
          BC059869.1 - 11 - 12 1819 6809310 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002757.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phactr3

          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          actin binding
          protein phosphatase inhibitor activity
          protein phosphatase 1 binding
          protein phosphatase type 1 regulator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001007154.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001007154.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001177789.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001177789.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001177790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001177790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001177791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_001177791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_028806.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 NM_028806.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022532.mm.1 TC0200002757.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline