Search
Search

Gabra4| Gene Symbol: Gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010251.2 | NP_034381.1 | 5 - 6 | 1079 | 67 | |
| GenBank mRNA | AK013727.1 | - | 5 - 6 | 1080 | 67 | |
| AK043806.1 | - | 5 - 6 | 697 | 67 | ||
| AK137354.1 | - | 5 - 6 | 696 | 67 | ||
| AK139970.1 | - | 5 - 6 | 831 | 67 | ||
| AK159299.1 | - | 5 - 6 | 1040 | 67 | ||
| AK162893.1 | - | 5 - 6 | 694 | 67 | ||
| BC094603.1 | - | 5 - 6 | 707 | 6828002 | 67 | |
Set Membership: |
|