Search
Search

Fstl5| Gene Symbol: Fstl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253719.1 | NP_001240648.1 | 5 - 6 | 779 | 91 | |
| NM_178673.4 | NP_848788.2 | 5 - 6 | 1039 | 91 | ||
| GenBank mRNA | AF374460.1 | - | 5 - 6 | 1013 | 91 | |
| AK033652.1 | - | 5 - 6 | 1030 | 91 | ||
| AK046070.1 | - | 5 - 6 | 780 | 91 | ||
| AK081152.1 | - | 5 - 6 | 1030 | 91 | ||
| AK138388.1 | - | 5 - 6 | 1331 | 91 | ||
| AK140611.1 | - | 5 - 6 | 1042 | 91 | ||
| BC075640.1 | - | 5 - 6 | 1002 | 30545529 | 91 | |
Set Membership: |
|