Search
Search

Ankrd2| Gene Symbol: Ankrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020033.1 | NP_064417.1 | 3 - 4 | 353 | 139 | |
| GenBank mRNA | AB076256.1 | - | 3 - 4 | 365 | 139 | |
| AJ245514.1 | - | 3 - 4 | 353 | 139 | ||
| AK132585.1 | - | 3 - 4 | 363 | 139 | ||
| BC138760.1 | - | 3 - 4 | 391 | 8861782 | 139 | |
| BC138761.1 | - | 3 - 4 | 391 | 8861783 | 139 | |
Set Membership: |
|