Search
Search

Acox3| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030721.2 | NP_109646.2 | 12 - 13 | 1483 | 113 | |
| GenBank mRNA | AK028818.1 | - | 12 - 13 | 1904 | 113 | |
| AK053960.1 | - | 12 - 13 | 1480 | 113 | ||
| AK128922.1 | - | 12 - 13 | 1443 | 113 | ||
| AK154112.1 | - | 11 - 12 | 1574 | 113 | ||
| AK171598.1 | - | 1466 | 113 | |||
| BC044725.2 | - | 12 - 13 | 1503 | 5374281 | 113 | |
| BC055019.1 | - | 12 - 13 | 1483 | 4162223 | 113 | |
Set Membership: |
|