Search
Search

Brcc3| Gene Symbol: Brcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166457.1 | NP_001159929.1 | 2 - 3 | 224 | 85 | |
| NM_001166459.1 | NP_001159931.1 | 2 - 3 | 224 | 85 | ||
| NM_145956.4 | NP_666068.1 | 2 - 3 | 224 | 85 | ||
| GenBank mRNA | AK011876.1 | - | 2 - 3 | 141 | 85 | |
| AK030736.1 | - | 2 - 3 | 154 | 85 | ||
| AK038919.1 | - | 2 - 3 | 142 | 85 | ||
| AK149844.1 | - | 2 - 3 | 156 | 85 | ||
| BC021313.1 | - | 2 - 3 | 171 | 5026715 | 85 | |
| BC048179.1 | - | 2 - 3 | 138 | 6825974 | 85 | |
| S68022.1 | - | 2 - 3 | 166 | 85 | ||
Set Membership: |
|