Search
Search

Tmprss2| Gene Symbol: Tmprss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015775.2 | NP_056590.2 | 7 - 8 | 911 | 131 | |
| GenBank mRNA | AF113596.1 | - | 7 - 8 | 781 | 131 | |
| AF199362.1 | - | 7 - 8 | 792 | 131 | ||
| AF243500.1 | - | 7 - 8 | 781 | 131 | ||
| AK141350.1 | - | 7 - 8 | 886 | 131 | ||
| AK146047.1 | - | 7 - 8 | 795 | 131 | ||
| AK161173.1 | - | 7 - 8 | 905 | 131 | ||
| BC038393.1 | - | 7 - 8 | 750 | 2648878 | 131 | |
| BC054348.1 | - | 7 - 8 | 911 | 2648889 | 131 | |
Set Membership: |
|