Search
Search

Chaf1b| Gene Symbol: Chaf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028083.4 | NP_082359.1 | 5 - 6 | 592 | 102 | |
| GenBank mRNA | AK011243.1 | - | 5 - 6 | 593 | 102 | |
| AK038378.1 | - | 5 - 6 | 562 | 102 | ||
| BC013532.1 | - | 4 - 5 | 502 | 4238801 | 102 | |
Set Membership: |
|