Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01215771_g1
          See other 1810059H22RIK GE Assays ›
          Gene Symbol:
          1810059H22Rik
          Gene Name:
          RIKEN cDNA 1810059H22 gene
          Gene Aliases:
          Gm577
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01215771_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810059H22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007907.1 - 1 - 2 556 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI646366.1 - 1 - 2 Chr. 5 - 148958598 - 148963996 [+] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 15411 Gm15411 OTTMUSG00000021229 Anti-sense Chr. 5 - 148953741 - 149015813 on NCBI Build GRCm38
          Downstream RIKEN cDNA 5930430L01 gene 5930430L01Rik 30.8kb Anti-sense Chr. 5 - 148990056 - 148995215 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2207342 AK144510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2207342 AI452232.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626231 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122135 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2207342 AK144510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2207342 AI452232.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122135 - -
          Whole Mouse Genome G4122A A_52_P422185 AK079377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P403578 AK007907.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04377268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04221533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04221530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04972319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04725468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24182450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440283_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432519_PM_at - -
          Mouse Genome 430 2.0 Array 1432519_at - -
          Mouse Genome 430 2.0 Array 1440283_at - -
          Mouse Genome 430 2.0 Array 1432519_PM_at - -
          Mouse Genome 430 2.0 Array 1440283_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69820_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69820_PM_at - -
          Mouse Expression Set 430 1432519_PM_at - -
          Mouse Expression Set 430 1432519_at - -
          Mouse Expression Set 430 1440283_at - -
          Mouse Expression Set 430 1440283_PM_at - -
          Murine Genome U74v2 Set 137567_PM_at - -
          Murine Genome U74v2 Set 137567_at - -
          Affymetrix HT Mouse Genome 430B Array 1432519_at - -
          Affymetrix HT Mouse Genome 430B Array 1440283_at - -
          Affymetrix HT Mouse Genome 430B Array 1440283_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432519_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358138 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline