Search
Search

Ripk2| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138952.3 | NP_620402.1 | 4 - 5 | 741 | 87 | |
| GenBank mRNA | AF461040.1 | - | 4 - 5 | 639 | 87 | |
| AF487539.1 | - | 4 - 5 | 639 | 87 | ||
| AK159593.1 | - | 4 - 5 | 765 | 87 | ||
| AK172319.1 | - | 4 - 5 | 763 | 87 | ||
| AK172417.1 | - | 4 - 5 | 763 | 87 | ||
| BC025611.1 | - | 2 - 3 | 259 | 87 | ||
| BC050845.1 | - | 4 - 5 | 754 | 87 | ||
| BC058088.1 | - | 4 - 5 | 886 | 87 | ||
| BC069878.1 | - | 4 - 5 | 766 | 5316066 | 87 | |
Set Membership: |
|