Search
Search

Cwh43| Gene Symbol: Cwh43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181323.2 | NP_851840.1 | 13 - 14 | 2079 | 72 | |
| GenBank mRNA | AK035550.1 | - | 12 - 13 | 1672 | 72 | |
| AK081976.1 | - | 13 - 14 | 2079 | 72 | ||
| BC016523.1 | - | 13 - 14 | 1907 | 3584006 | 72 | |
Set Membership: |
|