Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01216760_m1
          See other RNF6 GE Assays ›
          Gene Symbol:
          Rnf6
          Gene Name:
          ring finger protein (C3H2C3 type) 6
          Gene Aliases:
          1200013I08Rik, 5730419H05Rik, AA537053
          Chromosome Location:
          Chr.5: 146209193 - 146222191 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01216760_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256085.1 NP_001243014.1 3 - 4 311 133
          NM_001256086.1 NP_001243015.1 4 - 5 389 133
          NM_001256087.1 NP_001243016.1 3 - 4 443 133
          NM_028774.3 NP_083050.1 3 - 4 517 133
          GenBank mRNA AK004745.1 - 3 - 4 497 133
          AK029912.1 - 3 - 4 298 133
          AK150269.1 - 3 - 4 298 133
          AK152106.1 - 3 - 4 298 133
          AY039004.1 - 3 - 4 417 133
          BC138545.1 - 4 - 5 423 8861565 133
          BY136428.1 - 3 - 4 316 134
          CX237430.1 - 3 - 4 443 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003608.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf6

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          protein ubiquitination
          negative regulation of axon extension
          protein K27-linked ubiquitination
          positive regulation of transcription, DNA-templated
          regulation of androgen receptor signaling pathway
          protein K48-linked ubiquitination
          protein K6-linked ubiquitination
          DNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          androgen receptor binding
          ubiquitin protein ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_001256085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_001256085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_001256086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_001256086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_001256087.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_001256087.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_028774.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 NM_028774.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033121.mm.1 TC0500003608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_001256085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_001256085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_001256086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_001256086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_001256087.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_001256087.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_028774.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 NM_028774.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033123.mm.1 TC0500003608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500033126.mm.1 NM_001256086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033126.mm.1 NM_001256086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500033126.mm.1 TC0500003608.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline