Search
Search

Ddx58| Gene Symbol: Ddx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172689.3 | NP_766277.3 | 10 - 11 | 1613 | 74 | |
| GenBank mRNA | AK049305.1 | - | 1 - 2 | 204 | 74 | |
| AK052595.1 | - | 6 - 7 | 867 | 74 | ||
| AK128929.1 | - | 10 - 11 | 1548 | 74 | ||
| AK155930.1 | - | 10 - 11 | 1549 | 74 | ||
| AY553221.1 | - | 10 - 11 | 1489 | 74 | ||
| BC027369.1 | - | 10 - 11 | 1592 | 74 | ||
| BC106095.1 | - | 10 - 11 | 1533 | 74 | ||
| BC127163.1 | - | 10 - 11 | 1479 | 74 | ||
| BC152540.1 | - | 10 - 11 | 1481 | 74 | ||
Set Membership: |
|