Search
Search

Evi5l| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039578.3 | NP_001034667.3 | 7 - 8 | 1136 | 61 | |
| GenBank mRNA | AK045240.1 | - | 7 - 8 | 1137 | 61 | |
| BC042653.1 | - | 7 - 8 | 1116 | 61 | ||
Set Membership: |
|