Search
Search

Tnip2| Gene Symbol: Tnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139064.2 | NP_620703.1 | 3 - 4 | 735 | 98 | |
| GenBank mRNA | AJ304865.1 | - | 3 - 4 | 735 | 98 | |
| AK160165.1 | - | 3 - 4 | 693 | 98 | ||
| BC052083.1 | - | 4 - 5 | 781 | 6404610 | 98 | |
Set Membership: |
|