Search
Search

Nat8l| Gene Symbol: Nat8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001985.3 | NP_001001985.3 | 1 - 2 | 1226 | 58 | |
| GenBank mRNA | AK147307.1 | - | 1 - 2 | 1227 | 58 | |
| AK147594.1 | - | 1 - 2 | 505 | 58 | ||
| AK147703.1 | - | 1 - 2 | 564 | 58 | ||
| BC075616.1 | - | 1 - 2 | 307 | 58 | ||
| DQ174094.1 | - | 1 - 2 | 307 | 58 | ||
Set Membership: |
|