Search
Search

Myo16| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081397.1 | NP_001074866.1 | 14 - 15 | 1956 | 63 | |
| GenBank mRNA | AB429291.1 | - | 12 - 13 | 1564 | 63 | |
| AK082350.1 | - | 14 - 15 | 1692 | 63 | ||
| BC072580.1 | - | 14 - 15 | 1968 | 63 | ||
| BC082311.1 | - | 14 - 15 | 1822 | 63 | ||
| BC151049.1 | - | 14 - 15 | 1910 | 9087962 | 63 | |
| BC151051.1 | - | 14 - 15 | 1911 | 9087964 | 63 | |
| BC157966.1 | - | 14 - 15 | 1911 | 9007290 | 63 | |
| BC172122.1 | - | 14 - 15 | 1911 | 9054408 | 63 | |
Set Membership: |
|