Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218153_g1
          See other LRRC71 GE Assays ›
          Gene Symbol:
          Lrrc71
          Gene Name:
          leucine rich repeat containing 71
          Gene Aliases:
          4933430H15Rik
          Chromosome Location:
          Chr.3: 87736923 - 87748980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218153_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028971.1 NP_083247.1 14 - 15 1721 128
          GenBank mRNA AK016993.1 - 14 - 15 1649 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003233.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 AK220337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC054078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC072663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC079565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC079915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 AK220337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC054078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC072663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC079565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P485472 BC079915.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155159 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104008 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 AK220337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC054078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC072663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC079565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC079915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 AK220337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC054078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC072663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC079565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P485472 BC079915.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155159 - -
          Whole Mouse Genome G4122A A_52_P473542 AK220337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P473542 AK029938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P473542 NM_028971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P473542 AK016993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 AK220337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC054078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC072663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC079565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC079915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 AK220337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC054078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC072663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC079565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P485472 BC079915.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155159 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453786_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445223_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499200 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493009 - -
          Mouse Genome 430 2.0 Array 1446904_PM_at - -
          Mouse Genome 430 2.0 Array 1446904_at - -
          Mouse Genome 430 2.0 Array 1453786_PM_at - -
          Mouse Genome 430 2.0 Array 1445223_at - -
          Mouse Genome 430 2.0 Array 1445223_PM_at - -
          Mouse Genome 430 2.0 Array 1453786_at - -
          Affymetrix Mouse Clariom S Array PSR0300017169.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017169.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017169.mm.1 TC0300003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017170.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017170.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017170.mm.1 TC0300003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017167.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017167.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017167.mm.1 TC0300003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017166.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017166.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017166.mm.1 TC0300003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017168.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017168.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017168.mm.1 TC0300003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017164.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017164.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017164.mm.1 TC0300003233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017165.mm.1 NM_028971.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017165.mm.1 NM_028971.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017165.mm.1 TC0300003233.mm.1 MTA TC
          Mouse Expression Set 430 1453786_at - -
          Mouse Expression Set 430 1445223_at - -
          Mouse Expression Set 430 1453786_PM_at - -
          Mouse Expression Set 430 1445223_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453786_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453786_at - -
          Affymetrix HT Mouse Genome 430B Array 1445223_at - -
          Affymetrix HT Mouse Genome 430B Array 1445223_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 492772 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101170068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224605 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline