Search
Search

Ccser2| Gene Symbol: Ccser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027045.1 | NP_081321.1 | 5 - 6 | 2127 | 92 | |
| NM_028407.3 | NP_082683.2 | 3 - 4 | 299 | 92 | ||
| GenBank mRNA | AK005931.1 | - | 3 - 4 | 299 | 92 | |
| AK013689.1 | - | 5 - 6 | 2081 | 92 | ||
| AK129295.1 | - | 4 - 5 | 1699 | 92 | ||
| AK147383.1 | - | 5 - 6 | 2122 | 92 | ||
| AK172153.1 | - | 5 - 6 | 2101 | 92 | ||
| BC048641.1 | - | 3 - 4 | 298 | 92 | ||
| BC060947.1 | - | 3 - 4 | 298 | 92 | ||
| BC111465.1 | - | 4 - 5 | 508 | 92 | ||
| BC139090.1 | - | 4 - 5 | 1915 | 8862112 | 92 | |
Set Membership: |
|