Search
Search
| Gene Symbol: Phf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027949.1 | NP_082225.1 | 1 - 2 | 510 | 98 | |
| GenBank mRNA | AK005673.1 | - | 1 - 2 | 547 | 98 | |
| AK005852.1 | - | 1 - 2 | 510 | 98 | ||
| AK148191.1 | - | 1 - 2 | 593 | 98 | ||
| AK155352.1 | - | 1 - 2 | 311 | 98 | ||
| BC049113.1 | - | 1 - 2 | 607 | 98 | ||
| BC057174.1 | - | 1 - 2 | 357 | 5004450 | 98 | |
| BC132517.1 | - | 1 - 2 | 300 | 40130794 | 98 | |
| BC138571.1 | - | 1 - 2 | 300 | 8861591 | 98 | |
| BC145337.2 | - | 1 - 2 | 300 | 9053893 | 98 | |
| BC145338.1 | - | 1 - 2 | 300 | 98 | ||
Set Membership: |
|