Search
Search

Bod1l| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081422.2 | NP_001074891.1 | 22 - 23 | 8698 | 131 | |
| GenBank mRNA | AF013969.1 | - | 13 - 14 | 4157 | 131 | |
| AK122495.1 | - | 13 - 14 | 3842 | 131 | ||
| BC024569.1 | - | 1 - 2 | 48 | 131 | ||
Set Membership: |
|