Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01219058_g1
          See other TGFBR3L GE Assays ›
          Gene Symbol:
          Tgfbr3l
          Gene Name:
          transforming growth factor, beta receptor III-like
          Gene Aliases:
          Gm14378, OTTMUSG00000016477
          Chromosome Location:
          Chr.8: 4248214 - 4251423 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01219058_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfbr3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195258.1 NP_001182187.1 3 - 4 596 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003219.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976908 NM_001195258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976908 NM_001195258.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800000398.mm.1 NM_001195258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000398.mm.1 NM_001195258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000398.mm.1 TC0800003219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000402.mm.1 NM_001195258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000402.mm.1 NM_001195258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000402.mm.1 TC0800003219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000397.mm.1 NM_001195258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000397.mm.1 NM_001195258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000397.mm.1 TC0800003219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000399.mm.1 NM_001195258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000399.mm.1 NM_001195258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000399.mm.1 TC0800003219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000405.mm.1 NM_001195258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000405.mm.1 NM_001195258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000405.mm.1 TC0800003219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000400.mm.1 NM_001195258.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000400.mm.1 NM_001195258.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000400.mm.1 TC0800003219.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 822337 - -
          ABI Mouse Genome Survey Microarray 924384 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline