Search
Search

Fcrls| Gene Symbol: Fcrls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030707.3 | NP_109632.2 | 4 - 5 | 647 | 68 | |
| GenBank mRNA | AB028174.1 | - | 4 - 5 | 678 | 68 | |
| AF329486.1 | - | 4 - 5 | 648 | 68 | ||
| AK034125.1 | - | 4 - 5 | 648 | 68 | ||
| AK076123.1 | - | 4 - 5 | 681 | 68 | ||
Set Membership: |
|