Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165968.1 | NP_001159440.1 | 1 - 2 | 95 | 77 | |
NM_207263.2 | NP_997146.2 | 1 - 2 | 95 | 77 | ||
GenBank mRNA | AY518699.1 | - | 1 - 2 | 49 | 77 | |
BC120840.1 | - | 1 - 2 | 90 | 40129763 | 77 | |
BC144902.1 | - | 1 - 2 | 95 | 9053450 | 77 | |
BC144927.1 | - | 1 - 2 | 95 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |