| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500004646.mm.1 |
NM_019438.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004646.mm.1 |
NM_019438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004646.mm.1 |
TC0500000494.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004644.mm.1 |
NM_019438.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004644.mm.1 |
NM_019438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004644.mm.1 |
TC0500000494.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500004645.mm.1 |
NM_019438.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004645.mm.1 |
NM_019438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500004645.mm.1 |
TC0500000494.mm.1 |
MTA TC |