Search
Search
| Gene Symbol: Evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021292.2 | NP_067267.2 | 16 - 17 | 2535 | 61 | |
| GenBank mRNA | AK039759.1 | - | 16 - 17 | 2506 | 61 | |
| AK148262.1 | - | 16 - 17 | 2508 | 61 | ||
| AK161910.1 | - | 3 - 4 | 1185 | 61 | ||
Set Membership: |
|